Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC6A14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587998
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587998 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587998 Supplier Invitrogen™ Supplier No. PA587998
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YYNVIIAYSL YYMFASFQSE LPWKNCSSWS DKNCSRSPIV THCNVSTVNK GIQEIIQMNK SWVDINNFTC INGSEIYQPG QLPSEQYWNK VALQRSSGMN ETGV; Positive Samples: A-549, MCF7, SGC-7901; Cellular Location: Membrane, Multi-pass membrane protein.

This gene encodes a member of the solute carrier family 6. Members of this family are sodium and chloride dependent neurotransmitter transporters. The encoded protein transports both neutral and cationic amino acids.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC6A14
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.67 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene SLC6A14
Gene Accession No. Q9JMA9, Q9UN76
Gene Alias 1110007A17Rik; 9030613J17Rik; amino acid transporter ATB0+; amino acid transporter B0; amino acid transporter B0+; ATB(0+); Atb0+; ATB0plus; BMIQ11; carrier protein; CATB0plus; colonic system B0+ amino acid transporter; colonic system B0+ amino acid transporter CATB0+; LOW QUALITY PROTEIN: sodium- and chloride-dependent neutral and basic amino acid transporter B(0+); neutral cationic amino acid transporter B0,+; SLC6A14; Sodium- and chloride-dependent neutral and basic amino acid transporter B(0+); solute carrier family 6 (amino acid transporter), member 14; solute carrier family 6 (neurotransmitter transporter), member 14; solute carrier family 6 member 14
Gene Symbols SLC6A14
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11254, 56774
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.