Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC6A15 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587898
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587898 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587898 Supplier Invitrogen™ Supplier No. PA587898
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LVVFAVLGFK ANVINEKCIT QNSETIMKFL KMGNISQDII PHHINLSTVT AEDYHLVYDI IQKVKEEEFP ALHLNSCKIE EELNKAVQGT GLAFIAFTEA M; Positive Samples: Mouse kidney; Cellular Location: Membrane, Multi-pass membrane protein.

SLC6A15 (solute carrier family 6 (neutral amino acid transporter), member 15), also known as sodium-dependent neutral amino acid transporter B(0)AT2, transporter v7-3, NTT73 or sodium-coupled branched-chain amino-acid transporter 1 (SBAT1), is a 730 amino acid multi-pass membrane protein that acts as a sodium-dependent neutral amino acid transporter. A member of the sodium neurotransmitter symporter (SNF) family and SLC6A15 subfamily, SLC6A15 differs from other members of its family in that it does not appear to be chloride-dependent. SLC6A15 is expressed in brain and is encoded by a gene that maps to human chromosome 12, which encodes over 1,100 genes and comprises approximately 4. 5% of the human genome. Chromosome 12 is associated with a variety of diseases and afflictions, including hypochondrogenesis, achondrogenesis, Kniest dysplasia, Noonan syndrome and trisomy 12p, which causes facial developmental defects and seizure disorders.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC6A15
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.74 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Slc6a15
Gene Accession No. Q8BG16
Gene Alias AA536730; AI326450; AI326451; B0at2; B0AT2 protein; homolog of rat orphan transporter v7-3; hv7-3; NTT73; orphan sodium- and chloride-dependent neurotransmitter transporter NTT73; orphan transporter v7-3; SBAT1; SLC6A15; sodium- and chloride-dependent neurotransmitter transporter NTT73; sodium/chloride dependent neurotransmitter transporter Homo sapiens orphan neurotransmitter transporter NTT7; sodium-coupled branched-chain amino-acid transporter 1; Sodium-dependent neutral amino acid transporter B(0)AT2; solute carrier family 6 (neurotransmitter transporter), member 15; solute carrier family 6 (neutral amino acid transporter), member 15; solute carrier family 6 member 15; solute carrier family 6, member 15; transporter v7-3; V7-3
Gene Symbols Slc6a15
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 350-450 of human SLC6A15 (NP_8774991).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 103098
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.