Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC6A15 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501567818
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-7818 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-7818 Supplier Bethyl Laboratories, Inc Supplier No. A304475AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

SLC6A15 (solute carrier family 6 (neutral amino acid transporter), member 15), also known as sodium-dependent neutral amino acid transporter B(0)AT2, transporter v7-3, NTT73 or sodium-coupled branched-chain amino-acid transporter 1 (SBAT1), is a 730 amino acid multi-pass membrane protein that acts as a sodium-dependent neutral amino acid transporter. A member of the sodium neurotransmitter symporter (SNF) family and SLC6A15 subfamily, SLC6A15 differs from other members of its family in that it does not appear to be chloride-dependent. SLC6A15 is expressed in brain and is encoded by a gene that maps to human chromosome 12, which encodes over 1,100 genes and comprises approximately 4.5% of the human genome. Chromosome 12 is associated with a variety of diseases and afflictions, including hypochondrogenesis, achondrogenesis, Kniest dysplasia, Noo syndrome and trisomy 12p, which causes facial developmental defects and seizure disorders.

Specifications

Antigen SLC6A15
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene SLC6A15
Gene Accession No. Q9H2J7
Gene Alias Sodium-dependent neutral amino acid transporter B(0)AT2, Sodium-coupled branched-chain amino-acid transporter 1, Solute carrier family 6 member 15, Transporter v7-3, SLC6A15, B0AT2, NTT73, SBAT1
Gene Symbols SLC6A15
Host Species Rabbit
Immunogen Between 650 and 700
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55117
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.