Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC6A16 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP257364
Description
SLC6A16 Polyclonal specifically detects SLC6A16 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
SLC6A16 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence 1-4 ug/ml | |
NTT5solute carrier family 6 (neurotransmitter transporter), member 16, orphan sodium- and chloride-dependent neurotransmitter transporter NTT5, Solute carrier family 6 member 16, solute carrier family 6, member 16 | |
Rabbit | |
Affinity Purified | |
RUO | |
28968 | |
Human | |
IgG |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
SLC6A16 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:SQSFQFPVPWEKCPLTMNSSGFDPECERTTPSIYFWYQQALKASDRIEDGGSP | |
100 μL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction