Learn More
SLC6A16 Monoclonal Antibody (2E5)

Description
SLC6A16 Monoclonal Antibody for Western Blot, ICC/IF, ELISA
Specifications
Specifications
| Antigen | SLC6A16 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 2E5 |
| Concentration | 0.2-1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | SLC6A16 |
| Gene Accession No. | Q9GZN6 |
| Gene Alias | CTC-301O7.4; FLUP; FREQ; NT5; NTT5; Orphan sodium- and chloride-dependent neurotransmitter transporter NTT5; RGD1561338; Slc6a16; solute carrier family 6 (neurotransmitter transporter), member 16; solute carrier family 6 member 16; solute carrier family 6, member 16 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.