Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC6A16 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567437
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567437 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567437 Supplier Invitrogen™ Supplier No. PA567437
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SQSFQFPVPWE KCPLTMNSSG FDPECERTTP SIYFWYQQAL KASDRIEDGG SP Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 66%.

SLC6A16 shows structural characteristics of an Na(+)- and Cl(-)-dependent neurotransmitter transporter, including 12 transmembrane (TM) domains, intracellular N and C termini, and large extracellular loops containing multiple N-glycosylation sites.[supplied by OMIM, Mar 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC6A16
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC6A16
Gene Accession No. Q9GZN6
Gene Alias CTC-301O7.4; FLUP; FREQ; NT5; NTT5; Orphan sodium- and chloride-dependent neurotransmitter transporter NTT5; RGD1561338; Slc6a16; solute carrier family 6 (neurotransmitter transporter), member 16; solute carrier family 6 member 16; solute carrier family 6, member 16
Gene Symbols SLC6A16
Host Species Rabbit
Immunogen Recombinant Human SLC6A16. Recombinant protein control fragment (Product #RP-108099).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28968
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.