Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC6A17 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5139729
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5139729 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5139729 Supplier Invitrogen™ Supplier No. PA5139729
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: RHFHLLSDGS NTLSVSYKKG RMMKDISNLE ENDETRFILS KVPSEAPSPM PTHRSYLGPG STSPLETSGN PNGRYGSGYL LASTPESEL Highest antigen sequence indentity to the following orthologs: Rat - 97%, Mouse - 97%.

Functions as a sodium-dependent vesicular transporter selective for proline, glycine, leucine and alanine. In contrast to other members of this neurotransmitter transporter family, does not appear to be chloride-dependent.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC6A17
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC6A17
Gene Accession No. Q9H1V8
Gene Alias D130012J15Rik; MRT48; Ntt4; orphan sodium- and chloride-dependent neurotransmitter transporter NTT4; Rxt1; Slc6a17; Sodium-dependent neurotransmitter transporter NTT4; sodium-dependent neutral amino acid transporter SLC6A17; sodium-dependent orphan neurotramsmitter transporter NTT4; solute carrier family 6 (neurotransmitter transporter), member 17; solute carrier family 6 (neutral amino acid transporter), member 17; solute carrier family 6 member 17; solute carrier family 6, member 17
Gene Symbols SLC6A17
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC6A17. Recombinant protein control fragment (Product #RP-89587).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 388662
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.