Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC6A18 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104152
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104152 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104152 Supplier Invitrogen™ Supplier No. PA5104152
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SLC6A18.

The solute carrier (SLC) family, also known as the neurotransmitter transporter family, is one of the largest transporter families in the human genome. The SLC6 subgroup, which includes the dopamine transporter (DAT), the serotonin transporter (SERT) and the norepinephrine transporter (NET), contains sodium/chloride dependent, high-affinity plasma transport proteins. Sodium/chloride-dependent transporter XTRP2, also known as solute carrier family 6 member 18 (SLC6A18), is a 628 amino acid member of the SLC family. XTRP2 is composed of 12 exons, 11 introns and 8 tandem repeats. Highly expressed in the kidney, SLC6A18 is classified as an orphan transporter.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC6A18
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SLC6A18
Gene Accession No. Q96N87
Gene Alias b(0)AT3; B0AT3; D630001K16Rik; FLJ31236; Inactive sodium-dependent neutral amino acid transporter B(0)AT3; renal osmotic stress-induced Na-Cl organic solute cotransporter; Rosit; Slc6a18; sodium and chloride dependent transporter Xtrp2; sodium- and chloride-dependent transporter XTRP2; sodium channel-like protein; Sodium-dependent neutral amino acid transporter B(0)AT3; solute carrier family 6 (neurotransmitter transporter), member 18; solute carrier family 6 (neutral amino acid transporter), member 18; solute carrier family 6 member 18; solute carrier family 6, member 18; system B 0 neutral amino acid transporter AT3; system B(0) neutral amino acid transporter AT3; X transporter protein 2; Xt2; Xtrp2
Gene Symbols SLC6A18
Host Species Rabbit
Immunogen A synthesized peptide derived from human SLC6A18(Accession Q96N87), corresponding to amino acid residues I111-F161.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 348932
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.