Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC6A19 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA550828
Description
The antibody detects endogenous levels of total SLC6A19 protein.
This gene encodes a system B(0) transmembrane protein that actively transports most neutral amino acids across the apical membrane of epithelial cells. Mutations in this gene result in Hartnup disorder.
Specifications
SLC6A19 | |
Polyclonal | |
Unconjugated | |
SLC6A19 | |
B0AT1; HND; SLC6A19; sodium-dependent amino acid transporter system B0; sodium-dependent neutral amino acid transporter B(0)AT1; solute carrier family 6 (neurotransmitter transporter), member 19; solute carrier family 6 (neutral amino acid transporter), member 19; solute carrier family 6 member 19; System B(0) neutral amino acid transporter AT1; system B0 neutral amino acid transporter | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
340024 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1.5 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q695T7 | |
SLC6A19 | |
Synthetic peptide of human solute carrier family 6 (neutral amino acid transporter), member 19. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction