Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC6A19 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560276
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560276 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560276 Supplier Invitrogen™ Supplier No. PA560276
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MVRLVLPNPG LDARIPSLAE LETIEQEEAS SRPKWDNK Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 87%.

This gene encodes a system B(0) transmembrane protein that actively transports most neutral amino acids across the apical membrane of epithelial cells. Mutations in this gene result in Hartnup disorder.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC6A19
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC6A19
Gene Accession No. Q695T7
Gene Alias B0AT1; HND; SLC6A19; sodium-dependent amino acid transporter system B0; sodium-dependent neutral amino acid transporter B(0)AT1; solute carrier family 6 (neurotransmitter transporter), member 19; solute carrier family 6 (neutral amino acid transporter), member 19; solute carrier family 6 member 19; System B(0) neutral amino acid transporter AT1; system B0 neutral amino acid transporter
Gene Symbols SLC6A19
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC6A19. Recombinant protein control fragment (Product #RP-104115).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 340024
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.