Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC6A20 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568332
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568332 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568332 Supplier Invitrogen™ Supplier No. PA568332
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted to react with Mouse samples.

Mediates the calcium-dependent uptake of imino acids such as L-proline, N-methyl-L-proline and pipecolate as well as N-methylated amino acids. Involved in the transport of glycine.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC6A20
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SLC6A20
Gene Accession No. Q8VDB9, Q9NP91
Gene Alias A730081N20Rik; AU022428; IMINO-B; neurotransmitter transporter RB21A; orphan transporter XT3; rB21a; SIT1; Slc6a20; Slc6a20a; sodium- and chloride-dependent transporter XTRP3; Sodium- and chloride-dependent transporter XTRP3A; sodium/imino-acid transporter 1; solute carrier family 6 (neurotransmitter transporter), member 20; solute carrier family 6 (neurotransmitter transporter), member 20A; solute carrier family 6 (proline IMINO transporter), member 20; solute carrier family 6 member 20; Solute carrier family 6 member 20A; solute carrier family 6, member 20; transporter rB21A; Transporter rB21A homolog; X transporter protein 3; X transporter protein 3 similar 1; XT3; Xt3s1; Xtm3s1; Xtrp3; Xtrp3s1
Gene Symbols SLC6A20, Slc6a20a
Host Species Rabbit
Immunogen Synthesized peptide derived from human SLC6A20.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 102680, 54716
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.