Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC6A3/DAT1 Antibody (mAb16), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP222164UV
Description
SLC6A3/DAT1 Monoclonal specifically detects SLC6A3/DAT1 in Mouse, Rat, Human (Negative) samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
SLC6A3/DAT1 | |
Monoclonal | |
DyLight 350 | |
DA transporter, DATDAT1Solute carrier family 6 member 3, sodium-dependent dopamine transporter, solute carrier family 6 (neurotransmitter transporter, dopamine), member 3 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Mouse and rat. Little to no human reactivity has been observed. | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
mAb16 | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
SLC6A3 | |
Synthetic peptide corresponding to the N-terminus of rat DAT1. [UniProt# P23977] | |
0.1 mL | |
Neuroscience | |
6531 | |
Mouse, Rat, Human (Negative) | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction