Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC6A3 Rabbit anti-Human, Unconjugated, Clone: AACF-19, Abnova™

Catalog No. 89336386 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-336-386 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-336-386 Supplier Abnova Corporation Supplier No. MAB19986
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

SLC6A3 Monoclonal antibody specifically detects SLC6A3 in Human samples. It is validated for IP, WB-Ce, Flow Cyt.

Specifications

Antigen SLC6A3
Applications Immunoprecipitation, Western Blot, Flow Cytometry
Classification Monoclonal
Clone AACF-19
Conjugate Unconjugated
Description solute carrier family 6 (neurotransmitter transporter, dopamine), member 3
Gene SLC6A3
Gene Alias DAT, DAT1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to human SLC6A3
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 6531
Target Species Human
Content And Storage Store at -20°C for one year. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.