Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC6A4 Recombinant Rabbit Monoclonal Antibody (8Q4U2)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA551440
Description
Immunogen sequence: METTPLNSQK QLSACEDGED CQENGVLQKV VPTPGDKVES GQISNGYSAV PSPGAGDDTR HSIPATTTTL VAELHQGERE TWGK.
SERT is an integral membrane protein that transports the neurotransmitter serotonin from synaptic spaces into presynaptic neurons. The encoded protein terminates the action of serotonin and recycles it in a sodium-dependent manner. This protein is a target of psychomotor stimulants, such as amphetamines and cocaine, and is a member of the sodium:neurotransmitter symporter family. A repeat length polymorphism in the promoter of this gene has been shown to affect the rate of serotonin uptake and may play a role in sudden infant death syndrome, aggressive behavior in Alzheimer disease patients, and depression-susceptibility in people experiencing emotional trauma.
Specifications
SLC6A4 | |
Recombinant Monoclonal | |
1.08 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
P31645, P31652, Q60857 | |
SLC6A4 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-84 of human Serotonin transporter (NP_001036.1). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
Liquid |
ELISA, Western Blot | |
8Q4U2 | |
Unconjugated | |
SLC6A4 | |
5 OCD1; 5HT transporter; 5HTT; 5-HTT; 5-HTTLPR; 5-hydroxytryptamine (serotonin) transporter; AI323329; HGNC:11050; hSERT; HTT; Na+/Cl- dependent serotonin transporter; OCD1; OTTHUMP00000163633; serotonin transporter; serotonin transporter 1; Sert; SERT1; Slc6a4; Sodium-dependent serotonin transporter; Sodium-dependent serotonin transporter (5HT transporter) (5HTT); solute carrier family 6 (neurotransmitter transporter serotonin) member 4 (5-hydroxytryptamine (serotonin) transporter); Solute carrier family 6 (neurotransmitter transporter serotonin) member 4 (5-hydroxytryptamine (serotonin) transporter); solute carrier family 6 (neurotransmitter transporter), member 4; solute carrier family 6 (neurotransmitter transporter, serotonin), member 4; solute carrier family 6 member 4; solute carrier family 6, member 4 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
15567, 25553, 6532 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction