Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC6A7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550765
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550765 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550765 Supplier Invitrogen™ Supplier No. PA550765
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total SLC6A7 protein.

This gene is a member of the gamma-aminobutyric acid (GABA) neurotransmitter gene family and encodes a high-affinity mammalian brain L-proline transporter protein. This transporter protein differs from other sodium-dependent plasma membrane carriers by its pharmacological specificity, kinetic properties, and ionic requirements.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC6A7
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene SLC6A7
Gene Accession No. Q99884
Gene Alias AW455934; brain-specific L-proline transporter; L-proline transporter; proline transporter; Prot; Slc6a7; sodium-dependent proline transporter; solute carrier family 6 (neurotransmitter transporter), member 7; solute carrier family 6 (neurotransmitter transporter, L-proline), member 7; solute carrier family 6 member 7
Gene Symbols SLC6A7
Host Species Rabbit
Immunogen Synthetic peptide corresponding to a region derived from internal residues of human solute carrier family 6 (neurotransmitter transporter), member 7.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6534
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.