Learn More
Invitrogen™ SLC6A7 Polyclonal Antibody

Description
Immunogen sequence: SLTSDLPWEH CGNWWNTELC LEHRVSKDGN GALPLNLTCT VSPSEEYWSR YVLHIQGSQG IGSPG Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 89%.
Specifications
Specifications
| Antigen | SLC6A7 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | SLC6A7 |
| Gene Accession No. | Q99884 |
| Gene Alias | AW455934; brain-specific L-proline transporter; L-proline transporter; proline transporter; Prot; Slc6a7; sodium-dependent proline transporter; solute carrier family 6 (neurotransmitter transporter), member 7; solute carrier family 6 (neurotransmitter transporter, L-proline), member 7; solute carrier family 6 member 7 |
| Gene Symbols | SLC6A7 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.