Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC7A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590039
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590039 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590039 Supplier Invitrogen™ Supplier No. PA590039
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: Raji, Jurkat, 22RV1, Mouse testis, Rat brain, Rat liver; Cellular Location: Cell membrane, Multi-pass membrane protein Immunogen sequence: SLDADQARTP DGNLDQ.

SLC7A1 is a high-affinity, low capacity permease involved in the transport of the cationic amino acids (arginine, lysine and ornithine) in non-hepatic tissues. It may also function as an ecotropic retroviral leukemia receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC7A1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.42 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene SLC7A1
Gene Accession No. P30823, P30825, Q09143
Gene Alias 4831426K01Rik; AI447493; amino acid transporter, cationic 1; ATRC1; Atrc-1; Cat1; CAT-1; cationic amino acid transporter 1; cationic amino acid transporter, y+ system; cationic amino acid transporter-1; cationic amion acid transporter; y+ system; cationinc amino acid transporter 1; ecotropic retroviral leukemia receptor; Ecotropic retroviral leukemia receptor homolog; ecotropic retroviral receptor; ecotropic retrovirus receptor; Ecotropic retrovirus receptor homolog; ERR; HCAT1; high affinity cationic amino acid transporter 1; mCAT-1; Rec-1; REC1L; Rev-1; Slc7a1; solute carrier family 7 (cationic amino acid transporter, y+ system), member 1; solute carrier family 7 member 1; Solute carrier family 7 member A1 (amino acid transporter cationic 1); solute carrier family 7, member 1; system Y+ basic amino acid transporter
Gene Symbols SLC7A1
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11987, 25648, 6541
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.