Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC7A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542426
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542426 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542426 Supplier Invitrogen™ Supplier No. PA542426
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: LGFIMVSGFV KGSVKNWQLT EEDFGNTSGR LCLNNDTKEG KPGVGGFMPF Sequence homology: Cow: 100%; Horse: 93%; Human: 100%; Pig: 86%; Rabbit: 100%.

SLC7A1 is a high-affinity, low capacity permease involved in the transport of the cationic amino acids (arginine, lysine and ornithine) in non-hepatic tissues. It may also function as an ecotropic retroviral leukemia receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC7A1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SLC7A1
Gene Accession No. P30825
Gene Alias 4831426K01Rik; AI447493; amino acid transporter, cationic 1; ATRC1; Atrc-1; Cat1; CAT-1; cationic amino acid transporter 1; cationic amino acid transporter, y+ system; cationic amino acid transporter-1; cationic amion acid transporter; y+ system; cationinc amino acid transporter 1; ecotropic retroviral leukemia receptor; Ecotropic retroviral leukemia receptor homolog; ecotropic retroviral receptor; ecotropic retrovirus receptor; Ecotropic retrovirus receptor homolog; ERR; HCAT1; high affinity cationic amino acid transporter 1; mCAT-1; Rec-1; REC1L; Rev-1; Slc7a1; solute carrier family 7 (cationic amino acid transporter, y+ system), member 1; solute carrier family 7 member 1; Solute carrier family 7 member A1 (amino acid transporter cationic 1); solute carrier family 7, member 1; system Y+ basic amino acid transporter
Gene Symbols SLC7A1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human SLC7A1 (aa 201-250).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6541
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.