Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC7A10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563391
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563391 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563391 Supplier Invitrogen™ Supplier No. PA563391
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IFQGHFEELR PSNAFAFWMT P Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 90%.

SLC7A10, in association with 4F2HC (SLC3A2; MIM 158070), mediates high-affinity transport of D-serine and several other neutral amino acids.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC7A10
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC7A10
Gene Accession No. Q9NS82
Gene Alias AL024237; ASC1; Asc-1; Asc-type amino acid transporter 1; D- and L-serine transporter Asc-1; D7Bwg0847e; D-serine transporter; HASC-1; SLC7A10; solute carrier family 7 (cationic amino acid transporter, y+ system), member 10; solute carrier family 7 (neutral amino acid transporter light chain, asc system), member 10; solute carrier family 7 member 10; solute carrier family 7, (cationic amino acid transporter, y+ system) member 10; solute carrier family 7, (neutral amino acid transporter, y+ system) member 10
Gene Symbols SLC7A10
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC7A10. Recombinant protein control fragment (Product #RP-103287).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56301
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.