Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC7A14 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP159892
Description
SLC7A14 Polyclonal specifically detects SLC7A14 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
SLC7A14 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
FLJ37228, KIAA1613Solute carrier family 7 member 14, probable cationic amino acid transporter, solute carrier family 7 (cationic amino acid transporter, y+ system), member 14 | |
Rabbit | |
Protein A purified | |
RUO | |
57709 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1 mg/ml | |
Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
Q8TBB6 | |
SLC7A14 | |
Synthetic peptides corresponding to SLC7A14(solute carrier family 7 (cationic amino acid transporter, y+ system), member 14) The peptide sequence was selected from the N terminal of SLC7A14. Peptide sequence VAFFIGWNLILEYLIGTAAGASALSSMFDSLANHTISRWMADSVGTLNGL The peptide sequence for this immunogen was taken from within the described region. | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Chicken: 100%; Canine: 100%; Equine: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%. | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction