Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC7A14 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA561128
Description
Immunogen sequence: ADCEKEACSP VSEGDEFSGP ATNTCGAKNL PSLGDNEMLI GKSDKSTYNV NHPNYGTVDM TTGIEADESE NIYLIKLKKL IGPHYYTMRI RL Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.
The function remains unknown.
Specifications
SLC7A14 | |
Polyclonal | |
Unconjugated | |
SLC7A14 | |
A930013N06; BC061928; KIAA1613; PPP1R142; probable cationic amino acid transporter; protein phosphatase 1, regulatory subunit 142; SLC7A14; solute carrier family 7 (cationic amino acid transporter, y+ system), member 14; solute carrier family 7 (orphan transporter), member 14; solute carrier family 7 member 14; solute carrier family 7, member 14 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57709 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.32 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8TBB6 | |
SLC7A14 | |
Recombinant protein corresponding to Human SLC7A14. Recombinant protein control fragment (Product #RP-96253). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction