Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC7A14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542479
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542479 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542479 Supplier Invitrogen™ Supplier No. PA542479
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: VAFFIGWNLI LEYLIGTAAG ASALSSMFDS LANHTISRWM ADSVGTLNGL Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.

The function remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC7A14
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SLC7A14
Gene Accession No. Q8TBB6
Gene Alias A930013N06; BC061928; KIAA1613; PPP1R142; probable cationic amino acid transporter; protein phosphatase 1, regulatory subunit 142; SLC7A14; solute carrier family 7 (cationic amino acid transporter, y+ system), member 14; solute carrier family 7 (orphan transporter), member 14; solute carrier family 7 member 14; solute carrier family 7, member 14
Gene Symbols SLC7A14
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human SLC7A14 (aa 131-180).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57709
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.