Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC7A2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589910
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589910 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589910 Supplier Invitrogen™ Supplier No. PA589910
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PRNLSSPFIF HEKTSEF Positive Samples: 293T, HepG2, HeLa, Mouse liver, Mouse brain, Rat liver; Cellular Location: Cell membrane, Multi-pass membrane protein.

The protein encoded by this gene is a cationic amino acid transporter and a member of the APC (amino acid-polyamine-organocation) family of transporters. The encoded membrane protein is responsible for the cellular uptake of arginine, lysine and ornithine. Three transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC7A2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.68 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene Slc7a2
Gene Accession No. B5D5N9, P18581, P52569
Gene Alias 20.5; AI158848; amino acid transporter, cationic 2 (low affinity); ATRC2; CAT2; CAT-2; Cat2a; Cat2b; Cationic amino acid transporter 2; cationic amino acid transporter 2A; cationic amino acid transporter 2B; cationic amino acid transporter-2A; HCAT2; low affinity cationic amino acid transporter 2; Slc7a2; solute carrier family 7 (cationic amino acid transporter, y+ system), member 2; solute carrier family 7 member 2; solute carrier family 7, member 2; T-cell early activation protein; TEA
Gene Symbols Slc7a2
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11988, 64554, 6542
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.