Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC7A4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA539081
Description

Specifications
SLC7A4 | |
Polyclonal | |
Unconjugated | |
SLC7A4 | |
SLC7A4 | |
A synthetic peptide derived from the C-terminal region of human SLC7A4 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O43246 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6545 | |
-20°C | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction