Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC7A5/LAT1 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11039038
Description
SLC7A5/LAT1 Polyclonal specifically detects SLC7A5/LAT1 in Human, Canine, Monkey, Rabbit samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
SLC7A5/LAT1 | |
Polyclonal | |
Unconjugated | |
PBS with 0.1% Sodium Azide | |
SLC7A5 | |
Synthetic 14 amino acid peptide from N-terminus of human SLC7A5/LAT1. | |
0.05 mg | |
Primary | |
This is specific to the Slc7a5 protein. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1.0 mg/mL | |
Immunohistochemistry, Immunohistochemistry-Paraffin 20 ug/ml | |
4F2 LC, CD98, CD98LC, D16S469EL-type amino acid transporter 1, E16, Integral membrane protein E16, large neutral amino acids transporter 1, large neutral amino acids transporter small subunit 1, LAT1hLAT1, MPE16CD98 light chain, sodium-independent neutral amino acid transporter LAT1,4F2LC, solute carrier family 7 (cationic amino acid transporter, y+ system), member 5,4F2 light chain, Solute carrier family 7 member 5, y+ system cationic amino acid transporter | |
Rabbit | |
Affinity Purified | |
RUO | |
8140 | |
Human, Canine, Monkey, Rabbit | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction