Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC7A5/LAT1 Antibody (BU53) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$196.10
Specifications
Antigen | SLC7A5/LAT1 |
---|---|
Clone | BU53 |
Concentration | 1 mg/mL |
Dilution | Flow Cytometry 1 - 2 μg/mL, Immunohistochemistry 1:100 - 1:300, Immunohistochemistry-Paraffin 1:100 - 1:300, Flow (Cell Surface) 1 - 2 μg/mL, CyTOF-ready |
Applications | Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Flow Cytometry (Cell Surface), CyTOF |
Description
SLC7A5/LAT1 Monoclonal antibody specifically detects SLC7A5/LAT1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Flow (Cell Surface), CyTOFSpecifications
SLC7A5/LAT1 | |
1 mg/mL | |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Flow Cytometry (Cell Surface), CyTOF | |
Unconjugated | |
Mouse | |
Cancer | |
PBS | |
8140 | |
IgG2a | |
Protein G purified |
BU53 | |
Flow Cytometry 1 - 2 μg/mL, Immunohistochemistry 1:100 - 1:300, Immunohistochemistry-Paraffin 1:100 - 1:300, Flow (Cell Surface) 1 - 2 μg/mL, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Human | |
4F2 LC, CD98, CD98LC, D16S469EL-type amino acid transporter 1, E16, Integral membrane protein E16, large neutral amino acids transporter 1, large neutral amino acids transporter small subunit 1, LAT1hLAT1, MPE16CD98 light chain, sodium-independent neutral amino acid transporter LAT1,4F2LC, solute carrier family 7 (cationic amino acid transporter, y+ system), member 5,4F2 light chain, Solute carrier family 7 member 5, y+ system cationic amino acid transporter | |
Plasma cell line 8226 | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title