Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC7A5/LAT1 Antibody (EP3-1), DyLight 405 , Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP320115V
Description
SLC7A5/LAT1 Monoclonal antibody specifically detects SLC7A5/LAT1 in Human samples. It is validated for ImmunofluorescenceSpecifications
SLC7A5/LAT1 | |
Monoclonal | |
DyLight 405 | |
4F2 LC, CD98, CD98LC, D16S469EL-type amino acid transporter 1, E16, Integral membrane protein E16, large neutral amino acids transporter 1, large neutral amino acids transporter small subunit 1, LAT1hLAT1, MPE16CD98 light chain, sodium-independent neutral amino acid transporter LAT1,4F2LC, solute carrier family 7 (cationic amino acid transporter, y+ system), member 5,4F2 light chain, Solute carrier family 7 member 5, y+ system cationic amino acid transporter | |
Immunised mice and constructed IgG1 Fab phage display libraries from the spleen cells. SLC7A5/LAT1-reactive recombinant Fab fragments were retrieved by epitope peptide panning. | |
0.1 mL | |
Cancer | |
8140 | |
Store at 4°C in the dark. | |
IgG1 κ |
Immunofluorescence | |
EP3-1 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction