Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC7A5/LAT1 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP151935
Description
SLC7A5/LAT1 Polyclonal specifically detects SLC7A5/LAT1 in Human samples. It is validated for Western Blot, Peptide ELISA.Specifications
SLC7A5/LAT1 | |
Polyclonal | |
Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:64000 | |
NP_003477.4 | |
SLC7A5 | |
Peptide with sequence C-RHRKPELERPIKVN corresponding to internal region according to NP_003477.4. | |
0.1 mg | |
Cancer | |
8140 | |
Human | |
IgG |
Western Blot, ELISA | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
4F2 LC, CD98, CD98LC, D16S469EL-type amino acid transporter 1, E16, Integral membrane protein E16, large neutral amino acids transporter 1, large neutral amino acids transporter small subunit 1, LAT1hLAT1, MPE16CD98 light chain, sodium-independent neutral amino acid transporter LAT1,4F2LC, solute carrier family 7 (cationic amino acid transporter, y+ system), member 5,4F2 light chain, Solute carrier family 7 member 5, y+ system cationic amino acid transporter | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
Expected from sequence similarity: Dog, Mouse, Rat and Cow. | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction