Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC7A5/LAT1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
Specifications
Antigen | SLC7A5/LAT1 |
---|---|
Concentration | 1.9 mg/mL |
Dilution | Immunohistochemistry 5 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017809
![]() |
Novus Biologicals
NB1007340.025MG |
0.025 mg | N/A | N/A | N/A | ||||
Description
SLC7A5/LAT1 Polyclonal specifically detects SLC7A5/LAT1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
SLC7A5/LAT1 | |
Immunohistochemistry 5 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
Human | |
Q01650 | |
8140 | |
KLH conjugated synthetic peptide (RHRKPEL) representing aa 419-425 of Human LAT1. | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
1.9 mg/mL | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Cancer | |
PBS with No Preservative | |
4F2 LC, CD98, CD98LC, D16S469EL-type amino acid transporter 1, E16, Integral membrane protein E16, large neutral amino acids transporter 1, large neutral amino acids transporter small subunit 1, LAT1hLAT1, MPE16CD98 light chain, sodium-independent neutral amino acid transporter LAT1,4F2LC, solute carrier family 7 (cationic amino acid transporter, y+ system), member 5,4F2 light chain, Solute carrier family 7 member 5, y+ system cationic amino acid transporter | |
SLC7A5 | |
IgG | |
IgG purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title