Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC7A5 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS10125R

Description
Solute carrier family 7 member 5 also known as large neutral amino acids transporter small subunit 1 is a protein that in humans is encoded by the SLC7A5 gene.It is sodium-independent, high-affinity transport of large neutral amino acids such as phenylalanine, tyrosine, leucine, arginine and tryptophan, when associated with SLC3A2/4F2hc.It involved in cellular amino acid uptake and the transport of L-DOPA across the blood-brain barrier, and that of thyroid hormones triiodothyronine (T3) and thyroxine (T4) across the cell membrane in tissues such as placenta.Acts as an amino acid exchanger and plays a role in neuronal cell proliferation (neurogenesis) in brain.Specifications
SLC7A5 | |
Polyclonal | |
Unconjugated | |
SLC7A5 | |
4F2; 4F2 LC; 4F2 light chain; 4F2HC; 4F2LC; 4T2HC; CD98; CD98 light chain; CD98HC; CD98LC; D0H16S474E; D16S469E; E16; hLAT1; Integral membrane protein E16; large neutral amino acids transporter 1; Large neutral amino acids transporter small subunit 1; Lat1; LAT-1; L-type amino acid transporter 1; MDU1; MP hLAT1; MPE16; NACAE; Protein TA1; SLC7A5; sodium-independent neutral amino acid transporter LAT1; solute carrier family 7 (amino acid transporter light chain, L system), member 5; solute carrier family 7 (cationic amino acid transporter, y+ system), member 5; Solute carrier family 7 member 5; Ta1; tumor-associated protein 1; y+ system cationic amino acid transporter | |
Rabbit | |
Protein A | |
RUO | |
8140 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q01650 | |
SLC7A5 | |
KLH conjugated synthetic peptide derived from human SLC7A5. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction