Learn More
Invitrogen™ SLC7A5 Recombinant Rabbit Monoclonal Antibody (0P6E8)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA558039

Description
Immunogen sequence: FAAAKLLALA LIILLGFVQI GKGDVSNLDP NFSFEGTKLD VGNIVLALYS GLFAYGGWNY LNFVTEEMIN PYRNLPLAII ISLPIVTLVY VLTNLAYFTT L.
Solute carrier family 7 member 5 also known as large neutral amino acids transporter small subunit 1 is a protein that in humans is encoded by the SLC7A5 gene.It is sodium-independent, high-affinity transport of large neutral amino acids such as phenylalanine, tyrosine, leucine, arginine and tryptophan, when associated with SLC3A2/4F2hc.It involved in cellular amino acid uptake and the transport of L-DOPA across the blood-brain barrier, and that of thyroid hormones triiodothyronine (T3) and thyroxine (T4) across the cell membrane in tissues such as placenta.Acts as an amino acid exchanger and plays a role in neuronal cell proliferation (neurogenesis) in brain.
Specifications
SLC7A5 | |
Recombinant Monoclonal | |
0.7 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
Q01650 | |
SLC7A5 | |
A synthetic peptide corresponding to a sequence within amino acids 200-300 of human LAT1/SLC7A5 (NP_003477.4). | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA, Western Blot | |
0P6E8 | |
Unconjugated | |
SLC7A5 | |
4F2; 4F2 LC; 4F2 light chain; 4F2HC; 4F2LC; 4T2HC; CD98; CD98 light chain; CD98HC; CD98LC; D0H16S474E; D16S469E; E16; hLAT1; integral membrane protein E16; large neutral amino acids transporter 1; large neutral amino acids transporter small subunit 1; LAT1; LAT-1; L-type amino acid transporter 1; MDU1; MP hLAT1; MPE16; NACAE; Protein TA1; SLC7A5; sodium-independent neutral amino acid transporter LAT1; solute carrier family 7 (amino acid transporter light chain, L system), member 5; solute carrier family 7 (cationic amino acid transporter, y+ system), member 5; solute carrier family 7 member 5; Ta1; tumor-associated protein 1; y+ system cationic amino acid transporter | |
Rabbit | |
Protein A | |
RUO | |
8140 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.