Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC7A7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557574
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557574 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557574 Supplier Invitrogen™ Supplier No. PA557574
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DSTEYEVASQ PEVETSPLGD GASPGPEQVK LKK Highest antigen sequence identity to the following orthologs: Mouse - 68%, Rat - 58%.

The protein encoded by this gene is the light subunit of a cationic amino acid transporter. This sodium-independent transporter is formed when the light subunit encoded by this gene dimerizes with the heavy subunit transporter protein SLC3A2. This transporter is found in epithelial cell membranes where it transfers cationic and large neutral amino acids from the cell to the extracellular space. Defects in this gene are a cause of lysinuric protein intolerance (LPI). Several transcript variants encoding the same protein have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC7A7
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC7A7
Gene Accession No. Q9UM01
Gene Alias AI790233; amino acid transporter SLC7A7 y+LAT1; LAT3; LPI; monocyte amino acid permease 2; MOP-2; my+lat1; Slc7a7; solute carrier family 7 (amino acid transporter light chain, y+L system), member 7; solute carrier family 7 (cationic amino acid transporter, y+ system), member 7; solute carrier family 7 member 7; y(+)L-type amino acid transporter 1; Y+L amino acid transporter 1; Y+L amino acid transporter 1; solute carrier family 7 member 7; y+LAT1; y+LAT-1
Gene Symbols SLC7A7
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC7A7. Recombinant protein control fragment (Product #RP-95664).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9056
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.