Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC7A8 Antibody (OTI7D6), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274215H
Description
SLC7A8 Monoclonal specifically detects SLC7A8 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
SLC7A8 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
SLC7A8 | |
Full-length protein expressed in 293T cell transfected with human SLC7A8 expression vector. | |
0.1 mL | |
23428.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI7D6 | |
Western Blot, Immunohistochemistry | |
hLAT2, integral membrane protein E16H, large neutral amino acids transporter small subunit 2, LAT2L-type amino acid transporter 2, LPI-PC1, solute carrier family 7 (amino acid transporter, L-type), member 8, Solute carrier family 7 member 8, y+ system), member 8 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction