Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC7A8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563887
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563887 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563887 Supplier Invitrogen™ Supplier No. PA563887
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: WQHKPKCFSD FIELLTLVSQ KMCVVVYPEV ERGSGTEEAN ED Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 79%.

SLC7A8 is sodium-independent, high-affinity transport of large neutral amino acids. It has higher affinity for L-phenylalanine than LAT1 but lower affinity for glutamine and serine. L-alanine is transported at physiological concentrations. SLC7A8 also plays a role in basolateral (re)absorption of neutral amino acids.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC7A8
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC7A8
Gene Accession No. Q9UHI5
Gene Alias 4F2-LC5; 4F2LC-5; AA408822; cationic amino acid transporter y+ system; glycoprotein-associated amino acid transporter LAT2; hLAT2; integral membrane protein E16H; Large neutral amino acids transporter small subunit 2; LAT2; Lat4; LPI-PC1; L-type amino acid transporter 2; mLAT2; Slc7a8; solute carrier family 7 (amino acid transporter light chain, L system), member 8; solute carrier family 7 (amino acid transporter, L-type), member 8; solute carrier family 7 (cationic amino acid transporter, y+ system), member 8; solute carrier family 7 member 8; solute carrier family 8 (cationic amino acid transporter, y+ system), member 7
Gene Symbols SLC7A8
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC7A8. Recombinant protein control fragment (Product #RP-104269).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23428
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.