Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC7A9 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA550887
Description
The antibody detects endogenous levels of total SLC7A9 protein.
Involved in the high-affinity. sodium-independent transport of cystine and neutral and dibasic amino acids (system b(0.+)-like activity). Thought to be responsible for the high-affinity reabsorption of cystine in the kidney tubule.
Specifications
SLC7A9 | |
Polyclonal | |
Unconjugated | |
SLC7A9 | |
b(0,+)AT; b(0,+)AT1; B(0,+)-type amino acid transporter 1; b>o>, + amino acid transporter; b>o>, +AT; BAT1; CSNU3; FLJ94301; glycoprotein-associated amino acid transporter b0,+AT1; glycoprotein-associated amino acid transporter b0,+AT1; SLC7A9; solute carrier family 7 (amino acid transporter light chain, bo,+ system), member 9; solute carrier family 7 (cationic amino acid transporter, y+ system), member 9; solute carrier family 7 (glycoprotein-associated amino acid transporter light chain, bo,+ system), member 9; solute carrier family 7 member 9 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11136 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
2.4 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
P82251 | |
SLC7A9 | |
Synthetic peptide corresponding to residues near the N terminal of human solute carrier family 7 (amino acid transporter light chain, bo,+ system), member 9. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction