Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC7A9 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA560039
Description
Immunogen sequence: YKFGWAQKIS KPITMHLQML MEVVPPEEDP E Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 87%.
Involved in the high-affinity. sodium-independent transport of cystine and neutral and dibasic amino acids (system b(0.+)-like activity). Thought to be responsible for the high-affinity reabsorption of cystine in the kidney tubule.
Specifications
SLC7A9 | |
Polyclonal | |
Unconjugated | |
SLC7A9 | |
b(0,+)AT; b(0,+)AT1; B(0,+)-type amino acid transporter 1; b>o>, + amino acid transporter; b>o>, +AT; BAT1; CSNU3; FLJ94301; glycoprotein-associated amino acid transporter b0,+AT1; glycoprotein-associated amino acid transporter b0,+AT1; SLC7A9; solute carrier family 7 (amino acid transporter light chain, bo,+ system), member 9; solute carrier family 7 (cationic amino acid transporter, y+ system), member 9; solute carrier family 7 (glycoprotein-associated amino acid transporter light chain, bo,+ system), member 9; solute carrier family 7 member 9 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11136 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P82251 | |
SLC7A9 | |
Recombinant protein corresponding to Human SLC7A9. Recombinant protein control fragment (Product #RP-98858). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction