Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC7A9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144622
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144622 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144622 Supplier Invitrogen™ Supplier No. PA5144622
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLC7A9 Polyclonal Antibody for Western Blot

Involved in the high-affinity. sodium-independent transport of cystine and neutral and dibasic amino acids (system b(0.+)-like activity). Thought to be responsible for the high-affinity reabsorption of cystine in the kidney tubule.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC7A9
Applications Western Blot
Classification Polyclonal
Concentration 1.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300
Gene SLC7A9
Gene Accession No. P82251
Gene Alias b(0,+)AT; b(0,+)AT1; B(0,+)-type amino acid transporter 1; b>o>, + amino acid transporter; b>o>, +AT; BAT1; CSNU3; FLJ94301; glycoprotein-associated amino acid transporter b0,+AT1; glycoprotein-associated amino acid transporter b0,+AT1; SLC7A9; solute carrier family 7 (amino acid transporter light chain, bo,+ system), member 9; solute carrier family 7 (cationic amino acid transporter, y+ system), member 9; solute carrier family 7 (glycoprotein-associated amino acid transporter light chain, bo,+ system), member 9; solute carrier family 7 member 9
Gene Symbols SLC7A9
Host Species Rabbit
Immunogen Synthetic peptide encompassing a sequence within the Intracellular domain of human SLC7A9. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11136
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.