Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC8A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584833
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584833 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584833 Supplier Invitrogen™ Supplier No. PA584833
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RHAADQARKA VSMHEVNTEV TENDPVSKIF FEQGTYQCLE Highest antigen sequence indentity to the following orthologs: Rat - 90%, Mouse - 93%.

Ca2+ plays a critical role in intracellular signaling and thus regulation of calcium balance is critical for the maintenance of this signaling function. The plasma membrane Na+/Ca2+ (NaCX) exchangers mediate Na+-dependent Ca2+ efflux in a wide variety of cell types. NaCX can move Ca2+ either into or out of cells, depending on the net Na+, Ca2+, and K+ gradient across the membrane. In mammals, at least 5 distinct genes code for the exchangers: Three NaCX (NaCX-1, NaCX-2, and NaCX-3), and two in the NaCKX family (NaCKX-1 and NaCKX-2). NaCX-1 is most prominently expressed in the heart where it plays a major role in excitation-contraction coupling (Matsuoka et al., 1993). NACX-1 may also be a target for protection of the heart and brain from ischemia and reperfusion injury (Hagihara et al., 2005; Bano et al., 2005; Choi, 2005).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC8A1
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Slc8a1
Gene Accession No. P32418
Gene Alias AI852629; AV344025; CNC; D930008O12Rik; Na(+)/Ca(2+)-exchange protein 1; Na/Ca exchanger; Na+/Ca++ exchanger; Na+/Ca2+ exchanger; Na-Ca exchanger; Ncx; Ncx1; Slc8a1; Sodium/calcium exchanger 1; sodium/calcium exchanger isoform NaCa7; solute carrier family 8 (sodium/calcium exchanger), member 1; solute carrier family 8 member 1; solute carrier family 8 member A1; solute carrier family 8, member 1
Gene Symbols Slc8a1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC8A1. Recombinant protein control fragment (Product #RP-107729).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6546
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.