Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC8A2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562250
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562250 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562250 Supplier Invitrogen™ Supplier No. PA562250
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PANDSDTSTG GCQGSYRCQP GVLLPVWEPD DPSLGDKAA Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 95%.

Sodium/calcium exchanger proteins are integral membrane proteins primarily seen in cardiac cells. In cardiac myocytes, the concentration of Ca2+ alternates between low levels during relaxation and high levels during contraction. The Na+/Ca2+ exchanger 1 (NCX1) protein mediates Ca2+ extrusion from cardiac cells during relaxation. Four NCX1 isoforms (NCX1.1, NCX1.3, NCX1.7, and NCX1.10) result from alternate splicing. NCX1 mRNA is present at high levels in the heart, with lower levels present in the brain. NCX2 is most abundantly expressed in brain, in contrast the the broader distribution of NCX1, which is also expressed in heart, kidney, lung, smooth and skeletal muscle. The difference in expression for the transporter subtypes is believed to reflect differences in their functional roles. Regulation mechanisms for these exchanger proteins have not been fully characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC8A2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Slc8a2
Gene Accession No. Q9UPR5
Gene Alias KIAA1087; Na(+)/Ca(2+)-exchange protein 2; Na+/Ca2+-exchanging protein Nac2; NCX2; Slc8a2; sodium/calcium exchanger 2; sodium-calcium exchanger 2; solute carrier family 8 (sodium/calcium exchanger), member 2; solute carrier family 8 (sodium-calcium exchanger), member 2; solute carrier family 8 member 2; solute carrier family 8 member A2
Gene Symbols Slc8a2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC8A2. Recombinant protein control fragment (Product #RP-98990).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6543
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.