Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC8A3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550559
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550559 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550559 Supplier Invitrogen™ Supplier No. PA550559
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total SLC8A3 protein.

Rapidly transports Ca(2+) during excitation-contraction coupling. Ca(2+) is extruded from the cell during relaxation so as to prevent overloading of intracellular stores.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC8A3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 2.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene SLC8A3
Gene Accession No. P57103
Gene Alias AW742262; Na(+)/Ca(2+)-exchange protein 3; NCX3; NCX-3; Slc8a3; Sodium/calcium exchanger 3; sodium/calcium exchanger SLC8A3; solute carrier family 8 (sodium/calcium exchanger), member 3; solute carrier family 8 member 3; solute carrier family 8 member A3
Gene Symbols SLC8A3
Host Species Rabbit
Immunogen Synthetic peptide corresponding to a region derived from internal residues of human Solute carrier family 8 (sodium/calcium exchanger), member 3.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6547
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.