Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC8B1 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA5113781

Catalog No. PIPA5113781


Only null left
Add to Cart

Description

Description

Immunogen sequence: SIGDAFSDFT LARQGYPRMA FSACFGGIIF NILVGVGLGC LLQISRSHTE For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

SLC24A6 belongs to a family of potassium-dependent sodium/calcium exchangers that maintain cellular calcium homeostasis through the electrogenic counter transport of 4 sodium ions for 1 calcium ion and 1 potassium ion.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SLC8B1
Polyclonal
Unconjugated
SLC8B1
AF261233; Mitochondrial sodium/calcium exchanger protein; Na(+)/K(+)/Ca(2+)-exchange protein 6; Nckx6; NCLX; Slc24a6; Slc8b1; Sodium/calcium exchanger protein, mitochondrial; Sodium/potassium/calcium exchanger 6; sodium/potassium/calcium exchanger 6, mitochondrial; solute carrier family 24 (sodium/lithium/calcium exchanger), member 6; solute carrier family 24 (sodium/potassium/calcium exchanger), member 6; solute carrier family 24 member 6; solute carrier family 8 (sodium/lithium/calcium exchanger), member B1; solute carrier family 8 member B1
Rabbit
Affinity Chromatography
RUO
80024
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot, Surface Plasmon Resonance
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q6J4K2
SLC8B1
Synthetic peptide directed towards the middle region of human SLC24A6.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.