Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Slc9a10 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569291
Description
This target displays homology in the following species: Cow: 93%; Dog: 100%; Guinea Pig: 79%; Human: 100%; Mouse: 86%; Pig: 93%; Rabbit: 100%; Rat: 79%.
SLC9A10 is a member of the sodium-hydrogen exchanger (NHE) family (see SLC9A1, MIM 107310) and is required for male fertility and sperm motility (Wang et al., 2003 [PubMed 14634667]).
Specifications
Slc9a10 | |
Polyclonal | |
Unconjugated | |
SLC9C1 | |
Gm610; LOW QUALITY PROTEIN: sodium/hydrogen exchanger 10; Na(+)/H(+) exchanger 10; NHE; NHE-10; Slc9a10; SLC9C1; sNHE; Sodium/hydrogen exchanger 10; solute carrier family 10 member 10; Solute carrier family 9 member 10; Solute carrier family 9 member C1; solute carrier family 9, isoform 10; solute carrier family 9, member 10; solute carrier family 9, subfamily C (Na+-transporting carboxylic acid decarboxylase), member 1; spermNHE; sperm-NHE; Sperm-specific Na(+)/H(+) exchanger; sperm-specific sodium proton exchanger | |
Rabbit | |
Affinity Chromatography | |
RUO | |
288117 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
0 | |
SLC9C1 | |
synthetic peptide corresponding to a region of rat Slc9a10. | |
100 μL | |
Primary | |
Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction