Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC9A10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568333
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568333 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568333 Supplier Invitrogen™ Supplier No. PA568333
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLC9A10 Polyclonal Antibody for Western Blot

SLC9A10 is a member of the sodium-hydrogen exchanger (NHE) family (see SLC9A1, MIM 107310) and is required for male fertility and sperm motility (Wang et al., 2003 [PubMed 14634667]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC9A10
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SLC9C1
Gene Accession No. Q4G0N8
Gene Alias Gm610; LOW QUALITY PROTEIN: sodium/hydrogen exchanger 10; Na(+)/H(+) exchanger 10; NHE; NHE-10; Slc9a10; SLC9C1; sNHE; Sodium/hydrogen exchanger 10; solute carrier family 10 member 10; Solute carrier family 9 member 10; Solute carrier family 9 member C1; solute carrier family 9, isoform 10; solute carrier family 9, member 10; solute carrier family 9, subfamily C (Na+-transporting carboxylic acid decarboxylase), member 1; spermNHE; sperm-NHE; Sperm-specific Na(+)/H(+) exchanger; sperm-specific sodium proton exchanger
Gene Symbols SLC9C1
Host Species Rabbit
Immunogen Synthesized peptide derived from human SLC9A10.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285335
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.