Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC9A2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568335
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568335 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568335 Supplier Invitrogen™ Supplier No. PA568335
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted to react with Mouse and Rat samples.

Involved in pH regulation to eliminate acids generated by active metabolism or to counter adverse environmental conditions. Major proton extruding system driven by the inward sodium ion chemical gradient. Seems to play an important role in colonic sodium absorption.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC9A2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Slc9a2
Gene Accession No. P48763, Q3ZAS0, Q9UBY0
Gene Alias 2210416H12Rik; 4932415O19Rik; AV333884; H7; Na(+)/H(+) exchanger 2; Na/H ion exchanger; Nhe2; NHE-2; Slc9a2; Sodium/hydrogen exchanger 2; Solute carrier family 9 (sodium/hydrogen exchanger 2), antiporter 2, Na+/H+ (Na+/H+ exchanger 2); solute carrier family 9 (sodium/hydrogen exchanger), member 2; solute carrier family 9 member 2; solute carrier family 9 member A2; solute carrier family 9, member 2; solute carrier family 9, subfamily A (NHE2, cation proton antiporter 2), member 2
Gene Symbols Slc9a2
Host Species Rabbit
Immunogen Synthesized peptide derived from human SLC9A2.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 226999, 24783, 6549
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.