Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC9A4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557518
Description
Immunogen sequence: KQAIEMVETG ILSSTAFSIP HQAQRIQGIK RLSPEDVESI RDILTSNMYQ VRQRT Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 80%.

Specifications
SLC9A4 | |
Polyclonal | |
Unconjugated | |
SLC9A4 | |
Na(+)/H(+) exchanger 4; NHE4; NHE-4; SLC9A4; Sodium/hydrogen exchanger 4; solute carrier family 9 (sodium/hydrogen exchanger), isoform 4; solute carrier family 9 (sodium/hydrogen exchanger), member 4; solute carrier family 9 member 4; solute carrier family 9 member A4; solute carrier family 9, subfamily A (NHE4, cation proton antiporter 4), member 4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
389015 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.7 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6AI14 | |
SLC9A4 | |
Recombinant protein corresponding to Human SLC9A4. Recombinant protein control fragment (Product #RP-102695). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction