Learn More
Invitrogen™ SLC9A5 Polyclonal Antibody

Description
Immunogen sequence: GHVLSSTGLT LPSMPSRNSV AETSVTNLLR ESGSGACLDL QVIDTVRSGR DREDAVMHHL LCGGLYKPRR RYKASCSRHF ISEDAQERQD KEVFQQNMKR RL Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 99%.
Specifications
Specifications
| Antigen | SLC9A5 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | SLC9A5 |
| Gene Accession No. | Q14940 |
| Gene Alias | Gm696; membrane protein; Na(+)/H(+) exchanger 5; Nhe5; NHE-5; SLC9A5; sodium/hydrogen exchanger; sodium/hydrogen exchanger 5; sodium/hydrogen exchanger 5; solute carrier family 9 member A5; sodium/hydrogen exchanger isoform 5; solute carrier family 9 (sodium/hydrogen exchanger); solute carrier family 9 (sodium/hydrogen exchanger), isoform 5; solute carrier family 9 (sodium/hydrogen exchanger), member 5; solute carrier family 9 member 5; solute carrier family 9 member A5; solute carrier family 9, subfamily A (NHE5, cation proton antiporter 5), member 5 |
| Gene Symbols | SLC9A5 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.