Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC9A5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104162
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104162 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104162 Supplier Invitrogen™ Supplier No. PA5104162
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SLC9A5.

Na+/H+ exchangers-1-6 (Na+/H+ antiporters, NHE-1-6) are integral membrane proteins that are expressed in most mammalian tissues, where they regulate intracellular pH and cell volume. NHEs mediate the secondary active extrusion of hydrogen (H+) ions out of cells in exchange for extracellular sodium (Na+). Excluding NHE-1, which is ubiquitously expressed, NHE isoforms 2-6 have distinct tissue- and cell type-dependent expression and inhibitiory characteristics by amiloride analogs. Human NHE-5 transcript (solute carrier family 9, isoform-5, SLC9A5) is an 896 amino acid protein that is expressed in brain, testis, spleen and skeletal muscle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC9A5
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SLC9A5
Gene Accession No. Q14940, Q9Z0X2
Gene Alias Gm696; membrane protein; Na(+)/H(+) exchanger 5; Nhe5; NHE-5; SLC9A5; sodium/hydrogen exchanger; sodium/hydrogen exchanger 5; sodium/hydrogen exchanger 5; solute carrier family 9 member A5; sodium/hydrogen exchanger isoform 5; solute carrier family 9 (sodium/hydrogen exchanger); solute carrier family 9 (sodium/hydrogen exchanger), isoform 5; solute carrier family 9 (sodium/hydrogen exchanger), member 5; solute carrier family 9 member 5; solute carrier family 9 member A5; solute carrier family 9, subfamily A (NHE5, cation proton antiporter 5), member 5
Gene Symbols SLC9A5
Host Species Rabbit
Immunogen A synthesized peptide derived from human SLC9A5(Accession Q14940), corresponding to amino acid residues L815-L865.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 192215, 277973, 6553
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.