Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC9A6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101894
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101894 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101894 Supplier Invitrogen™ Supplier No. PA5101894
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SLC9A6.

Na+/H+ exchangers-1-6 (Na+/H+ antiporters, NHE-1-6) are integral membrane proteins that are expressed in most mammalian tissues where they regulate intracellular pH and cell volume. NHEs mediate the secondary active extrusion of hydrogen (H+) ions out of cells in exchange for extracellular sodium (Na+). Excluding NHE-1, which is ubiquitously expressed, the NHE isoforms 2-6 have distinct tissue- and cell type-dependent expression, and inhibitiory characteristics by amiloride analogs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC9A6
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SLC9A6
Gene Accession No. Q92581
Gene Alias 3732426M05; 6430520C02Rik; KIAA0267; mKIAA0267; MRSA; Na(+)/H(+) exchanger 6; NHE6; NHE-6; RGD1563582; SLC9A6; Sodium/hydrogen exchanger 6; solute carrier family 9 (sodium/hydrogen exchanger), isoform 6; solute carrier family 9 (sodium/hydrogen exchanger), member 6; Solute carrier family 9 member 6; solute carrier family 9 member A6; solute carrier family 9, subfamily A (NHE6, cation proton antiporter 6), member 6
Gene Symbols SLC9A6
Host Species Rabbit
Immunogen A synthesized peptide derived from human SLC9A6(Accession Q92581), corresponding to amino acid residues L633-A669.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10479, 236794
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.