Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC9A7 Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298612APC
Description
SLC9A7 Polyclonal antibody specifically detects SLC9A7 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
SLC9A7 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
Na(+)/H(+) exchanger 7, NHE-7, NHE7SLC9A6, nonselective sodium potassium/proton exchanger, sodium/hydrogen exchanger 7, solute carrier family 9 (sodium/hydrogen exchanger), solute carrier family 9 (sodium/hydrogen exchanger), isoform 7, solute carrier family 9 (sodium/hydrogen exchanger), member 7, Solute carrier family 9 member 7 | |
Produced in rabbits immunized with E. coli-derived Human SLC9A7 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin) | |
APC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
84679 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction