Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC9A7 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | SLC9A7 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
SLC9A7 Polyclonal specifically detects SLC9A7 in Mouse samples. It is validated for Western Blot.Specifications
SLC9A7 | |
Polyclonal | |
Rabbit | |
NP_796327 | |
84679 | |
The immunogen for this antibody is Slc9a7. Peptide sequence SSSYTASTSLECGRRTKSSSEEVLERDLGMGDQKVSSRGTPLVFPLQENA. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
Na(+)/H(+) exchanger 7, NHE-7, NHE7SLC9A6, nonselective sodium potassium/proton exchanger, sodium/hydrogen exchanger 7, solute carrier family 9 (sodium/hydrogen exchanger), solute carrier family 9 (sodium/hydrogen exchanger), isoform 7, solute carrier family 9 (sodium/hydrogen exchanger), member 7, Solute carrier family 9 member 7 | |
SLC9A7 | |
IgG | |
80 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title